Assay Details
| Gene Symbol: 4933400B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045432.1 | 3 - 4 | 467 | 112 | ||
| GenBank mRNA | AK016587.1 | - | 3 - 4 | 468 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | follicle stimulating hormone receptor | Fshr | -120.9kb | Anti-sense | Chr. 17 - 88984952 - 89200675 on NCBI Build GRCm38 | |
| Downstream | predicted gene 4719 | Gm4719 | 53.0kb | Sense | Chr. 17 - 89379241 - 89384993 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0600000178.rn
Set Membership: |
|

4933400B14Rik
Genomic Map