Assay Details
| Gene Symbol: 4930471L23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045460.1 | 1 - 2 | 172 | 199 | ||
| GenBank mRNA | AK015553.1 | - | 1 - 2 | 173 | 199 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elastin microfibril interfacer 2 | Emilin2 | FOAP-10 | -3.8kb | Anti-sense | Chr. 17 - 71252172 - 71311639 on NCBI Build GRCm38 |
| Downstream | ubiquitin specific peptidase 16 pseudogene | Gm4566 | 11.6kb | Anti-sense | Chr. 17 - 71333201 - 71341804 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930471L23Rik
Genomic Map