Assay Details
| Gene Symbol: 4930455J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045469.1 | 1 - 2 | 120 | 79 | ||
| GenBank mRNA | AK006099.1 | - | 1 - 2 | 124 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 28 (sodium-coupled nucleoside transporter), member 3 |
Slc28a3 | Cnt3 | -132.7kb | Sense | Chr. 13 - 58553307 - 58610877 on NCBI Build GRCm38 |
| Downstream |
neurotrophic tyrosine kinase, receptor, type 2 |
Ntrk2 |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
44.3kb | Anti-sense | Chr. 13 - 58806569 - 59133970 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2302000 | AK030052.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87535 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105959 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105959 | - | - |
| Whole Mouse Genome G4122A | A_52_P601221 | AK006099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P601221 | AK015470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P601215 | AK006099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P601215 | AK015470.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10922679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10821816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10534077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10473789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10856076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10562521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10770721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11060794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10729433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10533568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10760336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10704826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11090258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10839999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11060790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11012574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11076285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10473794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10997497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10997503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10797891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29074941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11090257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11090421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11012580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11076284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29075099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10690032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10856079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1430907_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430907_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430907_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013471.mm.1 | NR_045469.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013471.mm.1 | NR_045469.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013471.mm.1 | TC1300002104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013480.mm.1 | NR_045469.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013480.mm.1 | NR_045469.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013480.mm.1 | TC1300002104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013467.mm.1 | NR_045469.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013467.mm.1 | NR_045469.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013467.mm.1 | TC1300002104.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430907_PM_at | - | - |
| Mouse Expression Set 430 | 1430907_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430907_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430907_at | - | - |

4930455J16Rik
Genomic Map