Assay Details
| Gene Symbol: 1810007C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045472.1 | 2 - 3 | 472 | 77 | ||
| GenBank mRNA | AK007358.1 | - | 2 - 3 | 476 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 3110039M20 gene | 3110039M20Rik | -69.6kb | Anti-sense | Chr. 12 - 49389652 - 49407346 on NCBI Build GRCm38 | |
| Downstream | protein kinase D1 | Prkd1 |
PKD, PKD1, Pkcm, Prkcm |
863.1kb | Sense | Chr. 12 - 50341226 - 50649248 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1810007C17Rik
Genomic Map