Assay Details
| Gene Symbol: Ppp2r2cos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045505.1 | 2 - 3 | 1321 | 66 | ||
| GenBank mRNA | AK016607.1 | - | 2 - 3 | 1317 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 2, regulatory subunit B, gamma |
Ppp2r2c |
6330548O06Rik, IMYPNO, IMYPNO1, PR52 |
Anti-sense | Chr. 5 - 36866843 - 36955078 on NCBI Build GRCm38 | |
| Downstream | Wolfram syndrome 1 homolog (human) | Wfs1 |
AI481085, wolframin |
91.1kb | Sense | Chr. 5 - 36966104 - 36988982 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r2cos
Genomic Map