| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1444682_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1444682_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1444682_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1500009234.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009234.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009234.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009243.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009243.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009243.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009244.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009244.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009244.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009231.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009231.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009231.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009242.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009242.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009242.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009233.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009233.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009233.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500009235.mm.1 |
NR_028266.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009235.mm.1 |
NR_028266.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500009235.mm.1 |
TC1500001146.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1444682_PM_at |
- |
- |
| Mouse Expression Set 430 |
1444682_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1444682_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1444682_PM_at |
- |
- |