Assay Details
| Gene Symbol: 4933431G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045163.1 | 3 | 1113 | 105 | ||
| NR_045165.1 | 3 | 1517 | 105 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST3 beta-galactoside alpha-2,3-sialyltransferase 5 | St3gal5 |
3S-T, Siat9, [a]2 |
Anti-sense | Chr. 6 - 72097554 - 72154570 on NCBI Build GRCm38 | |
| Downstream | atonal homolog 8 (Drosophila) | Atoh8 |
4933425C05Rik, Hath6, Math6, bHLHa21, okadin |
84.5kb | Sense | Chr. 6 - 72206174 - 72235780 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933431G14Rik
Genomic Map