Assay Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045510.1 | 1 - 2 | 137 | 62 | ||
| GenBank mRNA | BB873716.1 | - | 1 - 2 | 121 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A930027H12 gene | A930027H12Rik | -461.5kb | Sense | Chr. 15 - 87097532 - 87254021 on NCBI Build GRCm38 | |
| Downstream | microRNA 6393 | Mir6393 |
Gm27424, mmu-mir-6393 |
75.2kb | Anti-sense | Chr. 15 - 87795850 - 87795943 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam19a5
Genomic Map