Assay Details
| Gene Symbol: Fam19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045514.1 | 5 - 6 | 2097 | 86 | ||
| GenBank mRNA | AK139403.1 | - | 5 - 6 | 2181 | 86 | |
| AK140763.1 | - | 5 - 6 | 2060 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4129 | Gm4129 | -530.5kb | Sense | Chr. 10 - 123196923 - 123199585 on NCBI Build GRCm38 | |
| Downstream |
eukaryotic translation initiation factor 3, subunit I pseudogene |
4930503E24Rik | 727.9kb | Sense | Chr. 10 - 124469074 - 124524488 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fam19a2
Genomic Map