Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281829.1 | NP_001268758.1 | 1 - 2 | 71 | 84 | |
| GenBank mRNA | AI182929.1 | - | 1 - 2 | 46 | 84 | |
| BC039982.1 | - | 1 - 2 | 66 | 84 | ||
| BY196857.1 | - | 1 - 2 | 73 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8A | Lrrc8a |
Lrrc8, mKIAA1437 |
-2.9kb | Sense | Chr. 2 - 30237769 - 30263790 on NCBI Build GRCm38 |
| Downstream | dolichol kinase | Dolk |
BC026973, Tmem15, mKIAA1094 |
17.3kb | Anti-sense | Chr. 2 - 30284229 - 30286354 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC039982.1,NM_001281829.1 transcripts.
Set Membership: |
|

Phyhd1
Genomic Map