Assay Details
| Gene Symbol: B4galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045570.1 | 5 - 6 | 1066 | 61 | ||
| GenBank mRNA | AK145250.1 | - | 5 - 6 | 1068 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 24 | Ccdc24 | Gm1028 | -4.8kb | Sense | Chr. 4 - 117869260 - 117872661 on NCBI Build GRCm38 |
| Downstream |
ATPase, H+ transporting, lysosomal V0 subunit B |
Atp6v0b |
2310024H13Rik, Atp6f, VMA16 |
6.5kb | Sense | Chr. 4 - 117884326 - 117887340 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

B4galt2
Genomic Map