Assay Details
| Gene Symbol: D17Ertd648e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045808.1 | 2 - 3 | 171 | 111 | ||
| GenBank mRNA | BG083152.2 | - | 2 - 3 | 171 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 28505 | Gm28505 | -772.3kb | Sense | Chr. 17 - 11076182 - 11090739 on NCBI Build GRCm38 | |
| Downstream |
1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta) |
Agpat4 | 1500003P24Rik | 254.4kb | Anti-sense | Chr. 17 - 12118709 - 12219649 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

D17Ertd648e
Genomic Map