Assay Details
Gene Symbol: Gm11201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045873.1 | 3 - 4 | 416 | 62 | ||
GenBank mRNA | AK154065.1 | - | 3 - 4 | 417 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
lectin, galactose binding, soluble 9 |
Lgals9 |
AA407335, AI194909, AI265545, LGALS35, Lgals5, gal-9, galectin-9 |
-43.2kb | Anti-sense | Chr. 11 - 78962979 - 78984924 on NCBI Build GRCm38 |
Downstream | WD repeat and SOCS box-containing 1 | Wsb1 |
1110056B13Rik, 2700038M07Rik |
208.6kb | Anti-sense | Chr. 11 - 79239382 - 79254700 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK154065.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
