Assay Details
| Gene Symbol: Gm10658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045886.1 | 2 - 3 | 891 | 107 | ||
| GenBank mRNA | AK143982.1 | - | 2 - 3 | 733 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | snurportin 1 | Snupn |
0610031A09Rik, Rnut1, Snupn1 |
-73.7kb | Anti-sense | Chr. 9 - 56950924 - 56983206 on NCBI Build GRCm38 |
| Downstream |
transcriptional regulator, SIN3A (yeast) |
Sin3a |
AW553200, Sin3, mKIAA4126, mSin3A |
14.2kb | Anti-sense | Chr. 9 - 57071991 - 57128368 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143982.1 transcripts.
Set Membership: |
|

Gm10658
Genomic Map