Assay Details
| Gene Symbol: 1700066N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045924.1 | 1 - 2 | 300 | 69 | ||
| GenBank mRNA | AK006908.1 | - | 1 - 2 | 301 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC051076 | BC051076 | Anti-sense | Chr. 5 - 87957622 - 87966352 on NCBI Build GRCm38 | ||
| Downstream |
calcium binding protein, spermatid specific 1 |
Cabs1 |
4931407G18Rik, AI428159, Clph |
100.0 | Anti-sense | Chr. 5 - 87979451 - 87981541 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700066N21Rik
Genomic Map