Assay Details
| Gene Symbol: 1700066N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045924.1 | 2 - 3 | 426 | 113 | ||
| GenBank mRNA | AK006908.1 | - | 2 - 3 | 427 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kappa | Csn3 |
AW208918, CSN10, Csnk |
Anti-sense | Chr. 5 - 87925575 - 87932545 on NCBI Build GRCm38 | |
| Downstream | cDNA sequence BC051076 | BC051076 | 2.5kb | Anti-sense | Chr. 5 - 87957622 - 87966352 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700066N21Rik
Genomic Map