Assay Details
| Gene Symbol: 1700066N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045924.1 | 3 - 4 | 497 | 149 | ||
| GenBank mRNA | AK006908.1 | - | 3 - 4 | 498 | 149 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC037156 | BC037156 | FDC-SP | Anti-sense | Chr. 5 - 87908779 - 87914171 on NCBI Build GRCm38 | |
| Downstream | casein kappa | Csn3 |
AW208918, CSN10, Csnk |
11.4kb | Anti-sense | Chr. 5 - 87925575 - 87932545 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700066N21Rik
Genomic Map