Assay Details
| Gene Symbol: Gm10677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046048.1 | 2 - 3 | 176 | 98 | ||
| GenBank mRNA | AK048734.1 | - | 2 - 3 | 177 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell adhesion molecule 1 | Cadm1 |
2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1 |
Anti-sense | Chr. 9 - 47530239 - 47853385 on NCBI Build GRCm38 | |
| Downstream |
neurexophilin and PC-esterase domain family, member 4 |
Nxpe4 |
C130036J11, D930028F11Rik, Fam55d |
348.0kb | Anti-sense | Chr. 9 - 48162001 - 48400025 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048734.1 transcripts.
Set Membership: |
|

Gm10677
Genomic Map