Assay Details
| Gene Symbol: Hoxd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073086.1 | 1 - 2 | 255 | 81 | ||
| GenBank mRNA | AK054090.1 | - | 1 - 2 | 256 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox D12 | Hoxd12 |
Hox-4.7, Hox-5.6 |
-1.9kb | Sense | Chr. 2 - 74675013 - 74677705 on NCBI Build GRCm38 |
| Downstream | homeobox D9 | Hoxd9 |
Hox-4.4, Hox-5.2 |
10.7kb | Sense | Chr. 2 - 74697763 - 74700208 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Hoxd11
Genomic Map