Assay Details
| Gene Symbol: Crem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110850.2 | NP_001104320.1 | 5 - 6 | 609 | 121 | |
| NM_001110854.1 | NP_001104324.2 | 3 - 4 | 251 | 121 | ||
| NM_001110855.1 | NP_001104325.2 | 2 - 3 | 215 | 121 | ||
| NM_001110856.2 | NP_001104326.1 | 7 - 8 | 792 | 121 | ||
| NM_001271504.1 | NP_001258433.1 | 6 - 7 | 756 | 121 | ||
| NM_001271505.1 | NP_001258434.1 | 7 - 8 | 834 | 121 | ||
| NM_001271506.1 | NP_001258435.1 | 7 - 8 | 945 | 121 | ||
| NM_013498.3 | NP_038526.2 | 6 - 7 | 645 | 121 | ||
| NR_073193.1 | 7 - 8 | 852 | 121 | |||
| GenBank mRNA | AJ292222.1 | - | 2 - 3 | 180 | 121 | |
| AJ311667.1 | - | 3 - 4 | 216 | 121 | ||
| AY738716.1 | - | 4 - 5 | 525 | 121 | ||
| AY738717.1 | - | 5 - 6 | 672 | 121 | ||
| AY738718.1 | - | 6 - 7 | 708 | 121 | ||
| AY738719.1 | - | 6 - 7 | 750 | 121 | ||
| AY738720.1 | - | 6 - 7 | 768 | 121 | ||
| AY738721.1 | - | 6 - 7 | 861 | 121 | ||
| BE689077.1 | - | 2 - 3 | 215 | 122 | ||
| M60285.1 | - | 6 - 7 | 573 | 121 | ||
| S64590.1 | - | 6 - 7 | 690 | 121 | ||
| S67786.1 | - | 2 - 3 | 202 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crem
Genomic Map