Assay Details
| Gene Symbol: Scarna8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028545.1 | 1 | 51 | 99 | ||
| GenBank mRNA | AF357402.1 | - | 1 | 51 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HAUS augmin-like complex, subunit 6 |
Haus6 |
6230416J20Rik, 9830144E06, D4Ertd27e, mKIAA1574 |
Sense | Chr. 4 - 86578855 - 86612076 on NCBI Build GRCm38 | |
| Downstream | perilipin 2 | Plin2 |
AA407157, ADPH, Adfp, Adrp |
40.3kb | Sense | Chr. 4 - 86626910 - 86670059 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scarna8
Genomic Map