Assay Details
| Gene Symbol: Crem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110850.2 | NP_001104320.1 | 2 - 3 | 223 | 135 | |
| NM_001110852.1 | NP_001104322.1 | 2 - 3 | 335 | 135 | ||
| NM_001110857.2 | NP_001104327.1 | 2 - 3 | 223 | 135 | ||
| NM_001110858.2 | NP_001104328.1 | 2 - 3 | 223 | 135 | ||
| NM_001271505.1 | NP_001258434.1 | 2 - 3 | 223 | 135 | ||
| NM_013498.3 | NP_038526.2 | 2 - 3 | 223 | 135 | ||
| GenBank mRNA | AI461618.1 | - | 3 - 4 | 332 | 135 | |
| AK009841.1 | - | 2 - 3 | 255 | 135 | ||
| AK010806.1 | - | 2 - 3 | 336 | 135 | ||
| AY738716.1 | - | 1 - 2 | 139 | 135 | ||
| AY738719.1 | - | 1 - 2 | 139 | 135 | ||
| BC034856.1 | - | 1 - 2 | 132 | 135 | ||
| M60285.1 | - | 2 - 3 | 151 | 135 | ||
More Information
Set Membership: |
|

Crem
Genomic Map