Assay Details
| Gene Symbol: Bin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270537.1 | NP_001257466.1 | 7 - 8 | 610 | 75 | |
| GenBank mRNA | CN691011.1 | - | 3 - 4 | 261 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bin2
Genomic Map