Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271498.1 | NP_001258427.1 | 3 - 4 | 676 | 84 | |
| GenBank mRNA | AK040940.1 | - | 3 - 4 | 528 | 84 | |
| AK080000.1 | - | 3 - 4 | 528 | 84 | ||
| AY221619.1 | - | 3 - 4 | 415 | 84 | ||
| BC145348.1 | - | 3 - 4 | 316 | 84 | ||
More Information
Set Membership: |
|

Il15ra
Genomic Map