Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291129.1 | NP_001278058.1 | 1 - 2 | 186 | 149 | |
| NM_001291130.1 | NP_001278059.1 | 1 - 2 | 186 | 100 | ||
| NM_010923.3 | NP_035053.1 | 1 - 2 | 186 | 154 | ||
| GenBank mRNA | AK003004.1 | - | 1 - 2 | 154 | 154 | |
| AK160626.1 | - | 1 - 2 | 154 | 154 | ||
| BC036984.1 | - | 1 - 2 | 144 | 4981517 | 154 | |
| GQ184462.1 | - | 1 - 2 | 141 | 100 | ||
| X83570.1 | - | 1 - 2 | 141 | 154 | ||
More Information
Set Membership: |
|

Nnat
Genomic Map