Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243769.1 | NP_001230698.1 | 2 - 3 | 262 | 91 | |
| NM_009861.3 | NP_033991.1 | 2 - 3 | 262 | 91 | ||
| GenBank mRNA | AK003098.1 | - | 2 - 3 | 183 | 91 | |
| AK075567.1 | - | 2 - 3 | 181 | 91 | ||
| AK144216.1 | - | 2 - 3 | 185 | 91 | ||
| AK150563.1 | - | 2 - 3 | 196 | 91 | ||
| AK151087.1 | - | 2 - 3 | 207 | 91 | ||
| AK151726.1 | - | 2 - 3 | 184 | 91 | ||
| AK153564.1 | - | 2 - 3 | 206 | 91 | ||
| AK154870.1 | - | 2 - 3 | 181 | 91 | ||
| AK159470.1 | - | 2 - 3 | 224 | 91 | ||
| AK166281.1 | - | 2 - 3 | 181 | 91 | ||
| AK167195.1 | - | 2 - 3 | 170 | 91 | ||
| AK167400.1 | - | 2 - 3 | 175 | 91 | ||
| AK167609.1 | - | 2 - 3 | 180 | 91 | ||
| AK168013.1 | - | 1 - 2 | 175 | 91 | ||
| AK168076.1 | - | 2 - 3 | 175 | 91 | ||
| AK168089.1 | - | 1 - 2 | 175 | 91 | ||
| AK168276.1 | - | 2 - 3 | 175 | 91 | ||
| AK168503.1 | - | 2 - 3 | 196 | 91 | ||
| AK168758.1 | - | 2 - 3 | 215 | 91 | ||
| AK168820.1 | - | 2 - 3 | 188 | 91 | ||
| AK169122.1 | - | 2 - 3 | 196 | 91 | ||
| AK169232.1 | - | 2 - 3 | 175 | 91 | ||
| AK169805.1 | - | 2 - 3 | 200 | 91 | ||
| BC064792.1 | - | 2 - 3 | 168 | 30105331 | 91 | |
| CD551112.1 | - | 2 - 3 | 249 | 91 | ||
| L11318.1 | - | 1 - 2 | 98 | 91 | ||
| L78075.1 | - | 1 - 2 | 102 | 91 | ||
| M79301.1 | - | 1 - 2 | 56 | 91 | ||
| U37720.1 | - | 2 - 3 | 170 | 91 | ||
More Information
Set Membership: |
|

Cdc42
Genomic Map