Assay Details
| Gene Symbol: Psip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290527.1 | NP_001277456.1 | 3 - 4 | 468 | 118 | |
| NM_133948.5 | NP_598709.1 | 3 - 4 | 468 | 118 | ||
| GenBank mRNA | AF339082.1 | - | 3 - 4 | 515 | 118 | |
| AF339083.1 | - | 3 - 4 | 515 | 118 | ||
| AJ308965.1 | - | 2 - 3 | 158 | 118 | ||
| AJ308966.1 | - | 2 - 3 | 158 | 118 | ||
| AK011572.1 | - | 3 - 4 | 448 | 118 | ||
| AK139598.1 | - | 2 - 3 | 567 | 118 | ||
| AK161426.1 | - | 3 - 4 | 442 | 118 | ||
| AK169600.1 | - | 2 - 3 | 249 | 118 | ||
| BC002104.1 | - | 3 - 4 | 414 | 118 | ||
| BC027215.1 | - | 3 - 4 | 515 | 118 | ||
| BC038820.1 | - | 2 - 3 | 217 | 118 | ||
| BC043079.1 | - | 3 - 4 | 415 | 6400529 | 118 | |
| BC052177.1 | - | 3 - 4 | 465 | 6512126 | 118 | |
| BY710824.1 | - | 3 - 4 | 447 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psip1
Genomic Map