Assay Details
| Gene Symbol: Arhgap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153820.3 | NP_722542.2 | 3 - 4 | 314 | 109 | |
| GenBank mRNA | AK018058.1 | - | 3 - 4 | 283 | 109 | |
| AK031022.1 | - | 3 - 4 | 270 | 109 | ||
| AK036520.1 | - | 3 - 4 | 315 | 109 | ||
| AK040796.1 | - | 3 - 4 | 306 | 109 | ||
| AK041323.1 | - | 3 - 4 | 307 | 109 | ||
| AK170537.1 | - | 3 - 4 | 312 | 109 | ||
| AK172407.1 | - | 3 - 4 | 305 | 109 | ||
| BC024887.1 | - | 3 - 4 | 268 | 3156334 | 109 | |
| BC034881.1 | - | 3 - 4 | 287 | 3468639 | 109 | |
More Information
Set Membership: |
|

Arhgap15
Genomic Map