Assay Details
| Gene Symbol: 4930506C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073374.1 | 1 - 2 | 102 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial pyruvate carrier 1 | Mpc1 |
0610006G08Rik, 3830411I18Rik, Brp44l |
-813.0 | Anti-sense | Chr. 17 - 8283813 - 8297667 on NCBI Build GRCm38 |
| Downstream | proline rich 18 | Prr18 | 9630019K15Rik | 29.3kb | Anti-sense | Chr. 17 - 8340406 - 8344113 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930506C21Rik
Genomic Map