Assay Details
| Gene Symbol: Runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073425.1 | 5 - 6 | 1565 | 71 | ||
| GenBank mRNA | AK134925.1 | - | 5 - 6 | 823 | 71 | |
| AK158925.1 | - | 5 - 6 | 1565 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride intracellular channel 5 | Clic5 |
5730531E12Rik, B330005L24, Gm322, jbg |
-215.8kb | Anti-sense | Chr. 17 - 44134678 - 44280168 on NCBI Build GRCm38 |
| Downstream |
runt related transcription factor 2, opposite strand 2 |
Runx2os2 | 9530072K05Rik | 62.7kb | Anti-sense | Chr. 17 - 44701390 - 44714098 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Runx2
Genomic Map