Assay Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073427.1 | 1 - 2 | 134 | 87 | ||
| GenBank mRNA | AK170778.1 | - | 1 - 2 | 113 | 87 | |
| BY238570.1 | - | 1 - 2 | 70 | 87 | ||
| CB318094.1 | - | 1 - 2 | 125 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 16984 |
Gm16984 | -9.8kb | Anti-sense | Chr. 13 - 34963853 - 34983318 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930529N20 gene | 4930529N20Rik | 173.0kb | Sense | Chr. 13 - 35167147 - 35167824 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170778.1 transcripts.
Set Membership: |
|

Eci2
Genomic Map