Assay Details
| Gene Symbol: Gm10364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073535.1 | 1 - 2 | 74 | 58 | ||
| GenBank mRNA | BC099969.1 | - | 1 - 2 | 74 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
neuroguidin, EIF4E binding protein |
Ngdn |
1500001L15Rik, Ngd, neuroguidin |
-14.3kb | Sense | Chr. 14 - 55015454 - 55024138 on NCBI Build GRCm38 |
| Downstream | zinc finger homeobox 2 | Zfhx2 | ZFH-5 | 17.3kb | Anti-sense | Chr. 14 - 55060265 - 55092433 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10364
Genomic Map