Assay Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_075099.1 | 2 - 3 | 421 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30505 |
Gm30505 | Anti-sense | Chr. 16 - 22874983 - 22894386 on NCBI Build GRCm38 | ||
| Downstream | fetuin beta | Fetub |
2310011O17Rik, AI255764, D17980, Gugu |
23.0kb | Sense | Chr. 16 - 22918382 - 22939768 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ahsg
Genomic Map