Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102282.1 | 1 - 2 | 495 | 75 | ||
| GenBank mRNA | DV056728.1 | - | 1 - 2 | 294 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 1, regulatory subunit 42 |
Ppp1r42 |
1700011J18Rik, 4930418G15Rik, Lrrc67, TLRR |
-21.4kb | Sense | Chr. 1 - 9968622 - 10013541 on NCBI Build GRCm38 |
| Downstream | centrosome and spindle pole associated protein 1 | Cspp1 |
2310020J12Rik, 4930413O22Rik, AA387550, BC055915, Cspp2, Cspp3 |
59.0 | Anti-sense | Chr. 1 - 10038218 - 10136771 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Cops5
Genomic Map