Assay Details
| Gene Symbol: 4933432K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102286.1 | 2 - 3 | 324 | 97 | ||
| GenBank mRNA | AK005578.1 | - | 2 - 3 | 323 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 2 | Hs3st2 |
6430516N12Rik, A830061E14Rik, AW491345 |
-85.6kb | Anti-sense | Chr. 7 - 121391610 - 121501768 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700025J12 gene | 1700025J12Rik | 449.0 | Anti-sense | Chr. 7 - 121589903 - 121591595 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933432K03Rik
Genomic Map