Assay Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102339.1 | 5 - 6 | 948 | 75 | ||
| GenBank mRNA | AK039466.1 | - | 6 - 7 | 855 | 75 | |
| BC065114.1 | - | 5 - 6 | 948 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 5 (iodide transporter), member 8 |
Slc5a8 |
Ait, SMCT |
-155.0kb | Anti-sense | Chr. 10 - 88885992 - 88929515 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 30769 |
Gm30769 | 168.9kb | Anti-sense | Chr. 10 - 89257155 - 89259142 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ano4
Genomic Map