Assay Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277273.1 | NP_001264202.1 | 13 - 14 | 1207 | 94 | |
| NM_025923.3 | NP_080199.1 | 13 - 14 | 1222 | 94 | ||
| NR_102382.1 | 11 - 12 | 989 | 94 | |||
| GenBank mRNA | AF513619.1 | - | 13 - 14 | 1222 | 94 | |
| AK008605.1 | - | 13 - 14 | 1108 | 94 | ||
| AK011520.1 | - | 13 - 14 | 1127 | 94 | ||
| AK011752.1 | - | 13 - 14 | 1099 | 94 | ||
| AK011903.1 | - | 13 - 14 | 1090 | 94 | ||
| AK018018.1 | - | 13 - 14 | 1104 | 94 | ||
| AK076017.1 | - | 13 - 14 | 1105 | 94 | ||
| AK168538.1 | - | 13 - 14 | 1106 | 94 | ||
| BC032876.1 | - | 13 - 14 | 1096 | 1282316 | 94 | |
| BC066181.1 | - | 13 - 14 | 1151 | 6435171 | 94 | |
More Information
Set Membership: |
|

Fancl
Genomic Map