| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001110783.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001277280.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001277281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001277284.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001277286.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_001277289.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NM_031158.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NR_102385.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
NR_102386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002254.mm.1 |
TC0800000291.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001110783.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001277280.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001277281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001277284.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001277286.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_001277289.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NM_031158.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NR_102385.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
NR_102386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002246.mm.1 |
TC0800000291.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001110783.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001277280.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001277281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001277284.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001277286.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_001277289.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NM_031158.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NR_102385.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
NR_102386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002252.mm.1 |
TC0800000291.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001110783.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001277280.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001277281.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001277284.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001277286.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_001277289.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NM_031158.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NR_102385.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
NR_102386.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002253.mm.1 |
TC0800000291.mm.1 |
MTA TC |