Assay Details
| Gene Symbol: Ank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102386.2 | 39 - 40 | 5060 | 58 | ||
| GenBank mRNA | AK147483.1 | - | 39 - 40 | 5060 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | K(lysine) acetyltransferase 6A | Kat6a |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
-188.2kb | Sense | Chr. 8 - 22859516 - 22943262 on NCBI Build GRCm38 |
| Downstream | predicted gene 15816 | Gm15816 | OTTMUSG00000026380 | 6.2kb | Anti-sense | Chr. 8 - 23138785 - 23149585 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Ank1
Genomic Map