Assay Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102387.1 | 13 - 14 | 2078 | 85 | ||
| GenBank mRNA | AY255674.1 | - | 13 - 14 | 1894 | 85 | |
| AY294283.1 | - | 13 - 14 | 1875 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
glutamate receptor interacting protein 1, opposite strand 1 |
Grip1os1 |
4930404E01Rik, 4930483C13Rik |
-202.2kb | Anti-sense | Chr. 10 - 119823658 - 119873140 on NCBI Build GRCm38 |
| Downstream | helicase (DNA) B | Helb |
AI447783, D10Ertd664e |
6.2kb | Anti-sense | Chr. 10 - 120083608 - 120112965 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Grip1
Genomic Map