Assay Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102387.1 | 14 - 15 | 2234 | 76 | ||
| GenBank mRNA | AY255674.1 | - | 14 - 15 | 2050 | 76 | |
| AY294283.1 | - | 14 - 15 | 2031 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | helicase (DNA) B | Helb |
AI447783, D10Ertd664e |
Anti-sense | Chr. 10 - 120083608 - 120112965 on NCBI Build GRCm38 | |
| Downstream | interleukin-1 receptor-associated kinase 3 | Irak3 |
4833428C18Rik, AI563835, IRAK-M |
54.4kb | Anti-sense | Chr. 10 - 120141648 - 120201597 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Grip1
Genomic Map