Assay Details
| Gene Symbol: Gm12992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102393.1 | 1 - 2 | 259 | 82 | ||
| NR_102394.1 | 1 - 2 | 259 | 82 | |||
| GenBank mRNA | BE655205.1 | - | 1 - 2 | 259 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/arginine-rich splicing factor 4 | Srsf4 |
5730499P16Rik, AW550192, MNCb-2616, SRp75, Sfrs4 |
-13.7kb | Anti-sense | Chr. 4 - 131873611 - 131901725 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 200B | Tmem200b | EG623230 | 1.2kb | Anti-sense | Chr. 4 - 131921200 - 131923956 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12992
Genomic Map