Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104374.1 | 1 - 2 | 66 | 75 | ||
| GenBank mRNA | BY252242.1 | - | 1 - 2 | 67 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | F-box protein 6 | Fbxo6 |
AA408845, FBG2, Fbs2, Fbx6b, Fbxo6b |
-6.4kb | Sense | Chr. 4 - 148145716 - 148152135 on NCBI Build GRCm38 |
| Downstream | F-box protein 2 | Fbxo2 |
FBG1, FBX2, Fbs1, Fbs2, NFB42, Prpl4 |
132.0 | Anti-sense | Chr. 4 - 148160226 - 148166417 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo44
Genomic Map