Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104484.1 | 19 - 20 | 3538 | 104 | ||
| GenBank mRNA | AF340028.1 | - | 19 - 20 | 3522 | 104 | |
| AK122437.1 | - | 8 - 9 | 901 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
wingless-type MMTV integration site family, member 5B |
Wnt5b |
AW545702, Wnt-5b |
-50.0kb | Sense | Chr. 6 - 119432531 - 119544824 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 3110021A11 gene | 3110021A11Rik | 229.3kb | Anti-sense | Chr. 6 - 119848193 - 119849029 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Erc1
Genomic Map