Assay Details
| Gene Symbol: Mir6337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105755.1 | 1 | 54 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 38, member 11 |
Slc38a11 | 9330158F14Rik | Sense | Chr. 2 - 65315799 - 65364394 on NCBI Build GRCm38 | |
| Downstream |
sodium channel, voltage-gated, type III, alpha |
Scn3a |
Gm1000, Nav1.3 |
92.7kb | Sense | Chr. 2 - 65457118 - 65567527 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6337
Genomic Map