Assay Details
| Gene Symbol: Mir6348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105766.1 | 1 | 53 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pre B cell leukemia homeobox 1 | Pbx1 |
2310056B04Rik, D230003C07Rik, Pbx-1 |
-58.4kb | Anti-sense | Chr. 1 - 168119364 - 168432264 on NCBI Build GRCm38 |
| Downstream | microRNA 6354 | Mir6354 |
Gm27282, mmu-mir-6354 |
528.2kb | Anti-sense | Chr. 1 - 169018974 - 169019089 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6348
Genomic Map