Assay Details
| Gene Symbol: Mir6481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105852.1 | 1 | 46 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12440 | Gm12440 | OTTMUSG00000006864 | -145.6kb | Anti-sense | Chr. 3 - 99049822 - 99053742 on NCBI Build GRCm38 |
| Downstream | T-box 15 | Tbx15 |
Tbx14, Tbx8, de |
40.9kb | Anti-sense | Chr. 3 - 99240347 - 99354260 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC0300002571.mm
This assay targets the same location as RTA transcript cluster(s) TC0200004301.rn
This assay targets the same location as RTA transcript cluster(s) TC0200004301.rn
Set Membership: |
|

Mir6481
Genomic Map