Assay Details
| Gene Symbol: Mir7001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105967.1 | 1 | 41 | 52 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13807 | Gm13807 | OTTMUSG00000014546 | -4.1kb | Anti-sense | Chr. 2 - 93403547 - 93417830 on NCBI Build GRCm38 |
| Downstream | predicted gene 10804 | Gm10804 | ENSMUSG00000075025 | 30.8kb | Anti-sense | Chr. 2 - 93452819 - 93469874 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7001
Genomic Map