Assay Details
| Gene Symbol: Mir7064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106031.1 | 1 | 48 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cysteinyl-tRNA synthetase | Cars | CA3 | Sense | Chr. 7 - 143557230 - 143600090 on NCBI Build GRCm38 | |
| Downstream |
tumor necrosis factor receptor superfamily, member 26 |
Tnfrsf26 |
Tnfrh3, Tnfrsf24 |
36.9kb | Sense | Chr. 7 - 143607685 - 143627845 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7064
Genomic Map