Assay Details
| Gene Symbol: Mir7082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106050.1 | 1 | 45 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38431 |
Gm38431 | Raver1-fdx1l | Sense | Chr. 9 - 21067514 - 21092008 on NCBI Build GRCm38 | |
| Downstream | tyrosine kinase 2 | Tyk2 | JTK1 | 28.5kb | Sense | Chr. 9 - 21104068 - 21131275 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7082
Genomic Map